siRNA Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292002.1 | NP_001278931.1 | 9 | 906 | ||
| NM_014487.5 | NP_055302.1 | 10 | 977 | |||
| XM_017008033.1 | XP_016863522.1 | 10 | 1121 | |||
| GenBank mRNA | AJ006591.2 | - | 10 | 922 | ||
| AK002106.1 | - | 10 | 926 | |||
| AK223609.1 | - | 10 | 928 | |||
| AK294089.1 | - | 9 | 860 | |||
| AK315463.1 | - | 10 | 963 | |||
| BC004421.1 | - | 10 | 953 | 3352916 | ||

ZNF330
Genomic Map