siRNA Details
| Gene Symbol: GOLIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308155.1 | NP_001295084.1 | 7, 8 | 1400 | ||
| NM_014498.4 | NP_055313.1 | 8, 9 | 1484 | |||
| XM_005247364.2 | XP_005247421.1 | 8, 9 | 1483 | |||
| XM_005247365.2 | XP_005247422.1 | 7, 8 | 1399 | |||
| XM_017006183.1 | XP_016861672.1 | 7, 8 | 879 | |||
| GenBank mRNA | AK307557.1 | - | 8, 9 | 1189 | ||
| AK307718.1 | - | 8, 9 | 1159 | |||
| U55853.1 | - | 8, 9 | 857 | |||

GOLIM4
Genomic Map