siRNA Details
| Gene Symbol: P2RY10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324218.1 | NP_001311147.1 | 3 | 1162 | ||
| NM_001324221.1 | NP_001311150.1 | 3 | 1068 | |||
| NM_001324225.1 | NP_001311154.1 | 4 | 1147 | |||
| NM_014499.3 | NP_055314.1 | 4 | 1211 | |||
| NM_198333.2 | NP_938147.1 | 2 | 1019 | |||
| GenBank mRNA | AK292138.1 | - | 4 | 1149 | ||
| AK313218.1 | - | 2 | 960 | |||
| BC051875.1 | - | 4 | 1122 | 5212240 | ||
| BC095498.1 | - | 1 | 847 | 6971933 | ||

P2RY10
Genomic Map