siRNA Details
| Gene Symbol: KCNMB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014505.5 | NP_055320.4 | 2 | 862 | ||
| XM_011538188.2 | XP_011536490.1 | 2 | 924 | |||
| GenBank mRNA | AF160967.1 | - | 2 | 746 | ||
| AF207992.1 | - | 2 | 409 | |||
| AF215891.1 | - | 2 | 464 | |||
| BC042446.2 | - | 2 | 571 | 3873518 | ||
| BC050621.1 | - | 2 | 503 | 5552807 | ||

KCNMB4
Genomic Map