siRNA Details
| Gene Symbol: MAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013283.4 | NP_037415.1 | 7 | 1078 | ||
| NM_182796.2 | NP_877725.1 | 7 | 1217 | |||
| GenBank mRNA | AB073390.1 | - | 5 | 2359 | ||
| AF113225.1 | - | 7 | 874 | |||
| AF182814.1 | - | 7 | 919 | |||
| AJ243721.1 | - | 7 | 1057 | |||
| AK096178.1 | - | 6 | 2338 | |||
| AK291919.1 | - | 7 | 1028 | |||
| AK294069.1 | - | 6 | 732 | |||
| AK312365.1 | - | 7 | 1027 | |||
| AL136664.1 | - | 7 | 991 | |||
| AY358695.1 | - | 7 | 972 | |||
| BC005218.1 | - | 7 | 998 | 3931953 | ||
| BC066645.1 | - | 7 | 953 | 5749043 | ||
| BC093030.1 | - | 7 | 987 | 30527606 | ||
| DQ395260.1 | - | 4 | 485 | |||
| DQ413182.1 | - | 3 | 483 | |||
| DQ413183.1 | - | 7 | 1005 | |||

MAT2B
Genomic Map