siRNA Details
| Gene Symbol: MAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013283.4 | NP_037415.1 | 7 | 1021 | ||
| NM_182796.2 | NP_877725.1 | 7 | 1160 | |||
| GenBank mRNA | AB073390.1 | - | 5 | 2302 | ||
| AF113225.1 | - | 7 | 817 | |||
| AF182814.1 | - | 7 | 862 | |||
| AJ243721.1 | - | 7 | 1000 | |||
| AK096178.1 | - | 6 | 2281 | |||
| AK291919.1 | - | 7 | 971 | |||
| AK294069.1 | - | 6 | 675 | |||
| AK312365.1 | - | 7 | 970 | |||
| AL136664.1 | - | 7 | 934 | |||
| AY358695.1 | - | 7 | 915 | |||
| BC005218.1 | - | 7 | 941 | 3931953 | ||
| BC066645.1 | - | 7 | 896 | 5749043 | ||
| BC093030.1 | - | 7 | 930 | 30527606 | ||
| DQ395260.1 | - | 4 | 428 | |||
| DQ413182.1 | - | 3 | 426 | |||

MAT2B
Genomic Map