siRNA Details
| Gene Symbol: CECR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017829.5 | NP_060299.4 | 7, 8 | 880 | ||
| NM_033070.2 | NP_149061.1 | 7, 8 | 955 | |||
| XM_005261236.3 | XP_005261293.1 | 7, 8 | 933 | |||
| XM_011546126.2 | XP_011544428.1 | 6, 7 | 751 | |||
| XM_011546127.1 | XP_011544429.1 | 7, 8 | 1370 | |||
| XM_017028781.1 | XP_016884270.1 | 7, 8 | 1422 | |||
| XM_017028782.1 | XP_016884271.1 | 8, 9 | 1749 | |||
| XM_017028783.1 | XP_016884272.1 | 8, 9 | 2186 | |||
| XM_017028784.1 | XP_016884273.1 | 8, 9 | 1749 | |||
| GenBank mRNA | AF273270.1 | - | 7, 8 | 880 | ||
| AF273271.1 | - | 7, 8 | 940 | |||
| AK000461.1 | - | 7, 8 | 953 | |||
| AK123646.1 | - | 2, 3 | 1436 | |||
| AK315156.1 | - | 7, 8 | 941 | |||
| AL833450.1 | - | 2, 3 | 2771 | |||
| BC042540.1 | - | 7, 8 | 939 | 4342582 | ||

CECR5
Genomic Map