siRNA Details
| Gene Symbol: SLCO1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019844.3 | NP_062818.1 | 16 | 2336 | ||
| GenBank mRNA | AF187815.1 | - | 14 | 2095 | ||
| AJ251506.1 | - | 14 | 2139 | |||
| AK055874.1 | - | 16 | 2314 | |||
| AY342017.1 | - | 14 | 2109 | |||

SLCO1B3
Genomic Map