siRNA Details
Gene Symbol: CDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271028.1 | NP_001257957.1 | 2 | 309 | ||
NM_021153.3 | NP_066976.1 | 2 | 309 | |||
NR_073130.1 | 2 | 309 | ||||
XM_011525931.2 | XP_011524233.1 | 2 | 412 | |||
XM_011525932.1 | XP_011524234.1 | 1 | 87 | |||
XM_017025711.1 | XP_016881200.1 | 2 | 334 | |||
GenBank mRNA | AJ007607.1 | - | 2 | 150 | ||
AK304000.1 | - | 2 | 259 | |||
AK315257.1 | - | 2 | 311 | |||
AY358654.1 | - | 1 | 48 |
