siRNA Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021643.3 | NP_067675.1 | 1 | 1512 | ||
| GenBank mRNA | AK313237.1 | - | 1 | 562 | ||
| AY245544.1 | - | 1 | 1495 | |||
| BC002637.2 | - | 1 | 562 | 3607549 | ||
| D87119.1 | - | 1 | 1301 | |||

TRIB2
Genomic Map