siRNA Details
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014028.3 | NP_054747.2 | 4 | 819 | ||
GenBank mRNA | AF077205.1 | - | 2 | 172 | ||
AF533891.1 | - | 4 | 803 | |||
AK075012.1 | - | 4 | 745 | |||
AK129854.1 | - | 4 | 813 | |||
AK314393.1 | - | 4 | 495 | |||
AY358795.1 | - | 4 | 810 | |||
BC068581.1 | - | 4 | 801 | 5298450 |
