siRNA Details
| Gene Symbol: OSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014028.3 | NP_054747.2 | 4 | 848 | ||
| GenBank mRNA | AF077205.1 | - | 2 | 201 | ||
| AF533891.1 | - | 4 | 832 | |||
| AK075012.1 | - | 4 | 774 | |||
| AK129854.1 | - | 4 | 842 | |||
| AK314393.1 | - | 4 | 524 | |||
| AY358795.1 | - | 4 | 839 | |||
| BC068581.1 | - | 4 | 830 | 5298450 | ||

OSTM1
Genomic Map