siRNA Details
| Gene Symbol: SLC27A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017372.2 | NP_001017372.1 | 10 | 2783 | ||
| NM_001317984.1 | NP_001304913.1 | 11 | 2607 | |||
| NM_014031.4 | NP_054750.1 | 11 | 2558 | |||
| GenBank mRNA | AA614135.1 | - | 1 | 436 | ||
| AF064254.1 | - | 10 | 1925 | |||
| AK313456.1 | - | 11 | 2206 | |||
| AL833117.1 | - | 10 | 2355 | |||
| BC041945.1 | - | 10 | 2782 | 5301306 | ||
| CR457129.1 | - | 10 | 1772 | |||

SLC27A6
Genomic Map