siRNA Details
| Gene Symbol: ATAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014109.3 | NP_054828.2 | 19 | 2726 | ||
| XM_011516994.2 | XP_011515296.1 | 19 | 3253 | |||
| XM_011516995.2 | XP_011515297.1 | 19 | 3253 | |||
| XM_011516996.2 | XP_011515298.1 | 8 | 1146 | |||
| GenBank mRNA | AF118088.1 | - | 2 | 184 | ||
| AK097133.1 | - | 19 | 2708 | |||
| AL833653.1 | - | 19 | 2480 | |||
| AY598335.1 | - | 19 | 2618 | |||
| BC010686.1 | - | 4 | 594 | |||
| BC113656.1 | - | 19 | 2640 | 8322708 | ||
| CR749832.1 | - | 20 | 2955 | |||

ATAD2
Genomic Map