siRNA Details
| Gene Symbol: ATAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014109.3 | NP_054828.2 | 25 | 3679 | ||
| XM_011516996.2 | XP_011515298.1 | 14 | 2099 | |||
| GenBank mRNA | AF118088.1 | - | 8 | 1137 | ||
| AL833653.1 | - | 25 | 3433 | |||
| AY598335.1 | - | 25 | 3571 | |||
| BC010686.1 | - | 10 | 1547 | |||
| BC019909.1 | - | 7 | 883 | |||
| BC113656.1 | - | 25 | 3593 | 8322708 | ||
| CR749832.1 | - | 26 | 3908 | |||

ATAD2
Genomic Map