siRNA Details
| Gene Symbol: UBE2T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014176.3 | NP_054895.1 | 2, 3 | 245 | ||
| GenBank mRNA | AB032931.1 | - | 1, 2 | 151 | ||
| AF160215.1 | - | 2, 3 | 202 | |||
| AF161499.1 | - | 2, 3 | 235 | |||
| AK000504.1 | - | 2, 3 | 200 | |||
| AY542309.1 | - | 1, 2 | 110 | |||
| BC004152.2 | - | 2, 3 | 207 | 2821806 | ||
| BC019284.1 | - | 2, 3 | 207 | 2821806 | ||
| BG776416.1 | - | 2, 3 | 246 | |||
| BP244265.1 | - | 2, 3 | 245 | |||

UBE2T
Genomic Map