siRNA Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014990.2 | NP_001014990.1 | 4, 5 | 267 | ||
| NM_014180.3 | NP_054899.2 | 5, 6 | 385 | |||
| GenBank mRNA | AF161507.1 | - | 5, 6 | 353 | ||
| AK000601.1 | - | 5, 6 | 372 | |||
| AK097880.1 | - | 1, 2 | 2022 | |||
| AK302784.1 | - | 4, 5 | 247 | |||
| BC012565.1 | - | 5, 6 | 355 | 4333117 | ||
| BM662504.1 | - | 2, 3 | 359 | |||
| BP372420.1 | - | 5, 6 | 385 | |||
| BU943770.1 | - | 5, 6 | 378 | |||
| CR933679.1 | - | 4, 5 | 246 | |||

MRPL22
Genomic Map