siRNA Details
| Gene Symbol: CFAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013242.2 | NP_037374.1 | 4 | 678 | ||
| NR_136632.1 | 4 | 711 | ||||
| GenBank mRNA | AF087845.1 | - | 4 | 658 | ||
| AF093680.1 | - | 4 | 657 | |||
| AK027099.1 | - | 4 | 599 | |||
| AK297819.1 | - | 4 | 633 | |||
| BC005152.2 | - | 4 | 647 | 3528301 | ||
| BU730963.1 | - | 3 | 633 | |||
| CN364433.1 | - | 4 | 638 | |||

CFAP20
Genomic Map