siRNA Details
| Gene Symbol: TBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013254.3 | NP_037386.1 | 4 | 485 | ||
| XM_005268809.1 | XP_005268866.1 | 4 | 563 | |||
| XM_005268810.1 | XP_005268867.1 | 4 | 545 | |||
| GenBank mRNA | AF174536.1 | - | 3 | 326 | ||
| AF191838.1 | - | 4 | 388 | |||
| AK291039.1 | - | 4 | 430 | |||
| BC034950.2 | - | 4 | 425 | 4824988 | ||
| DA788357.1 | - | 4 | 486 | |||

TBK1
Genomic Map