siRNA Details
| Gene Symbol: PRSS50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013270.4 | NP_037402.1 | 5 | 966 | ||
| GenBank mRNA | AF100707.1 | - | 5 | 966 | ||
| BC033016.1 | - | 5 | 957 | 4822455 | ||
| BC037775.2 | - | 5 | 928 | 5272458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS50| Gene Symbol: PRSS50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013270.4 | NP_037402.1 | 5 | 966 | ||
| GenBank mRNA | AF100707.1 | - | 5 | 966 | ||
| BC033016.1 | - | 5 | 957 | 4822455 | ||
| BC037775.2 | - | 5 | 928 | 5272458 | ||