siRNA Details
| Gene Symbol: TMOD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013353.2 | NP_037485.2 | 2 | 293 | ||
| XM_011509449.1 | XP_011507751.1 | 2 | 175 | |||
| XM_017001089.1 | XP_016856578.1 | 1 | 910 | |||
| XM_017001090.1 | XP_016856579.1 | 2 | 222 | |||
| GenBank mRNA | AF165217.1 | - | 2 | 170 | ||
| AF177173.2 | - | 2 | 238 | |||
| AK092700.1 | - | 2 | 175 | |||
| AK300675.1 | - | 2 | 203 | |||
| AK313280.1 | - | 2 | 202 | |||
| BC017810.1 | - | 2 | 203 | 4288769 | ||
| BX647346.1 | - | 2 | 203 | |||
| DC385872.1 | - | 2 | 293 | |||

TMOD4
Genomic Map