siRNA Details
| Gene Symbol: UBN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079514.2 | NP_001072982.1 | 12, 13 | 2323 | ||
| NM_001288656.1 | NP_001275585.1 | 11, 12 | 1757 | |||
| XM_005255277.3 | XP_005255334.1 | 12, 13 | 2508 | |||
| XM_005255278.3 | XP_005255335.1 | 12, 13 | 3472 | |||
| XM_011522465.1 | XP_011520767.1 | 12, 13 | 2508 | |||
| XM_011522466.2 | XP_011520768.1 | 7, 8 | 1149 | |||
| XM_017023189.1 | XP_016878678.1 | 12, 13 | 2508 | |||
| XM_017023191.1 | XP_016878680.1 | 5, 6 | 881 | |||
| GenBank mRNA | AF108460.1 | - | 12, 13 | 1818 | ||
| AF108461.1 | - | 11, 12 | 2405 | |||
| AK300131.1 | - | 11, 12 | 1757 | |||
| U19346.1 | - | 6, 7 | 663 | |||

UBN1
Genomic Map