siRNA Details
| Gene Symbol: SAC3D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013299.3 | NP_037431.3 | 2 | 1384 | ||
| GenBank mRNA | U79266.1 | - | 2 | 1379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAC3D1| Gene Symbol: SAC3D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013299.3 | NP_037431.3 | 2 | 1384 | ||
| GenBank mRNA | U79266.1 | - | 2 | 1379 | ||