siRNA Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013300.2 | NP_037432.2 | 3 | 658 | ||
| XM_005253875.4 | XP_005253932.1 | 3 | 168 | |||
| XM_011538246.2 | XP_011536548.1 | 3 | 186 | |||
| GenBank mRNA | AK225731.1 | - | 3 | 649 | ||
| AK296280.1 | - | 3 | 670 | |||
| AK297684.1 | - | 3 | 168 | |||
| AK308802.1 | - | 3 | 271 | |||
| BC020967.1 | - | 3 | 474 | 3848037 | ||
| CR457001.1 | - | 3 | 209 | |||
| U79274.1 | - | 3 | 625 | |||

FAM216A
Genomic Map