siRNA Details
Gene Symbol: C18orf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276342.1 | NP_001263271.1 | 7 | 789 | ||
NM_001318707.1 | NP_001305636.1 | 6 | 704 | |||
NM_001318708.1 | NP_001305637.1 | 8 | 848 | |||
NM_001318709.1 | NP_001305638.1 | 7 | 789 | |||
NM_013326.4 | NP_037458.3 | 9 | 933 | |||
XM_011525939.1 | XP_011524241.1 | 8 | 805 | |||
XM_017025717.1 | XP_016881206.1 | 7 | 742 | |||
GenBank mRNA | AF143536.1 | - | 8 | 680 | ||
AK057192.1 | - | 9 | 894 | |||
AK294169.1 | - | 6 | 651 | |||
AK295246.1 | - | 8 | 805 | |||
AK296230.1 | - | 7 | 740 | |||
BC002950.1 | - | 9 | 858 | 3538017 | ||
BC008305.1 | - | 9 | 865 | 3537084 |
