siRNA Details
| Gene Symbol: SEC61A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013336.3 | NP_037468.1 | 1, 2 | 196 | ||
| GenBank mRNA | AF077032.1 | - | 1, 2 | 102 | ||
| AF084458.1 | - | 1, 2 | 102 | |||
| AF346602.1 | - | 1, 2 | 78 | |||
| AK027423.1 | - | 1, 2 | 196 | |||
| AK074706.1 | - | 1, 2 | 196 | |||
| AL832821.1 | - | 1, 2 | 129 | |||

SEC61A1
Genomic Map