siRNA Details
| Gene Symbol: ALG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013339.3 | NP_037471.2 | 14 | 1464 | ||
| GenBank mRNA | AF063604.2 | - | 14 | 1419 | ||
| AF102851.1 | - | 14 | 1397 | |||
| AI479334.1 | - | 2 | 573 | |||
| AK022700.1 | - | 14 | 1402 | |||
| AK295286.1 | - | 6 | 554 | |||
| BC001253.1 | - | 14 | 1396 | 3452687 | ||
| CB215801.1 | - | 6 | 390 | |||

ALG6
Genomic Map