siRNA Details
| Gene Symbol: PKN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317926.1 | NP_001304855.1 | 11, 12 | 1894 | ||
| NM_013355.4 | NP_037487.2 | 11, 12 | 1894 | |||
| XM_005251946.3 | XP_005252003.1 | 11, 12 | 2449 | |||
| XM_006717080.2 | XP_006717143.1 | 11, 12 | 1963 | |||
| XM_017014649.1 | XP_016870138.1 | 11, 12 | 1965 | |||
| XM_017014650.1 | XP_016870139.1 | 11, 12 | 2103 | |||
| GenBank mRNA | AB019692.1 | - | 11, 12 | 1482 | ||
| BC062558.1 | - | 11, 12 | 1497 | 6649806 | ||

PKN3
Genomic Map