siRNA Details
| Gene Symbol: PURG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015508.2 | NP_001015508.1 | 1 | 1231 | ||
| NM_001323311.1 | NP_001310240.1 | 2 | 421 | |||
| NM_001323312.1 | NP_001310241.1 | 2 | 421 | |||
| NM_013357.2 | NP_037489.1 | 1 | 317 | |||
| GenBank mRNA | AF195513.2 | - | 1 | 1231 | ||
| AY077841.1 | - | 1 | 1231 | |||
| BC106708.2 | - | 1 | 317 | 40005076 | ||
| DR003356.1 | - | 2 | 426 | |||

PURG
Genomic Map