siRNA Details
| Gene Symbol: PADI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013358.2 | NP_037490.2 | 15 | 1739 | ||
| XM_017001101.1 | XP_016856590.1 | 15 | 1642 | |||
| XM_017001102.1 | XP_016856591.1 | 15 | 1642 | |||
| GenBank mRNA | AB033768.2 | - | 15 | 1730 | ||
| AK026652.1 | - | 15 | 1741 | |||
| AK127515.1 | - | 1 | 1018 | |||
| AK225541.1 | - | 15 | 1741 | |||
| AK293275.1 | - | 5 | 493 | |||
| AK298892.1 | - | 4 | 273 | |||
| AK314644.1 | - | 15 | 1740 | |||
| BC130574.1 | - | 15 | 1718 | 40146605 | ||
| BC136402.1 | - | 15 | 1718 | 9020389 | ||

PADI1
Genomic Map