siRNA Details
| Gene Symbol: SERTAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013376.3 | NP_037508.2 | 2 | 686 | ||
| GenBank mRNA | AF117959.1 | - | 2 | 657 | ||
| AF366402.1 | - | 2 | 638 | |||
| AK074652.1 | - | 2 | 686 | |||
| AK222970.1 | - | 2 | 653 | |||
| AK223011.1 | - | 2 | 653 | |||
| BC002670.2 | - | 2 | 635 | 3613213 | ||

SERTAD1
Genomic Map