siRNA Details
| Gene Symbol: SLC25A24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013386.4 | NP_037518.3 | 9 | 1330 | ||
| NM_213651.2 | NP_998816.1 | 9 | 1241 | |||
| XM_017001104.1 | XP_016856593.1 | 5 | 586 | |||
| GenBank mRNA | AF123303.1 | - | 9 | 1089 | ||
| AJ619961.1 | - | 9 | 1173 | |||
| AJ619987.1 | - | 9 | 1119 | |||
| AK292567.1 | - | 9 | 1241 | |||
| AK304165.1 | - | 9 | 1324 | |||
| AK316304.1 | - | 9 | 1284 | |||
| AK316506.1 | - | 9 | 1152 | |||
| BC014519.1 | - | 9 | 1186 | 3842039 | ||
| BC068561.1 | - | 9 | 1219 | 5259336 | ||

SLC25A24
Genomic Map