siRNA Details
| Gene Symbol: UBQLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013444.3 | NP_038472.2 | 1 | 824 | ||
| GenBank mRNA | AF189009.1 | - | 1 | 778 | ||
| AF293385.1 | - | 1 | 543 | |||
| AK297252.1 | - | 1 | 790 | |||
| AK302885.1 | - | 1 | 790 | |||
| AL442081.1 | - | 1 | 437 | |||
| BC069237.1 | - | 2 | 695 | 4543266 | ||

UBQLN2
Genomic Map