siRNA Details
| Gene Symbol: SLC2A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271711.1 | NP_001258640.1 | 3, 4 | 528 | ||
| NM_014580.4 | NP_055395.2 | 3, 4 | 528 | |||
| XM_006717084.3 | XP_006717147.1 | 3, 4 | 523 | |||
| XM_011518602.2 | XP_011516904.1 | 3, 4 | 522 | |||
| GenBank mRNA | AF289587.1 | - | 3, 4 | 435 | ||
| AJ245937.1 | - | 3, 4 | 438 | |||
| BC019043.2 | - | 3, 4 | 480 | 4641145 | ||
| HY025817.1 | - | 3, 4 | 393 | |||
| Y17801.1 | - | 3, 4 | 461 | |||

SLC2A8
Genomic Map