siRNA Details
| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199583.2 | NP_001186512.1 | 6 | 888 | ||
| NM_020804.4 | NP_065855.1 | 6 | 901 | |||
| XM_011514541.1 | XP_011512843.1 | 6 | 756 | |||
| GenBank mRNA | AB037800.2 | - | 6 | 858 | ||
| AF242529.1 | - | 6 | 824 | |||
| AK289956.1 | - | 6 | 886 | |||
| AK294132.1 | - | 6 | 888 | |||
| AL834211.1 | - | 6 | 834 | |||
| BC040228.1 | - | 6 | 873 | 5173129 | ||
| BC128219.1 | - | 3 | 281 | |||

PACSIN1
Genomic Map