siRNA Details
| Gene Symbol: FSCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020369.2 | NP_065102.1 | 2 | 550 | ||
| GenBank mRNA | AF281049.1 | - | 2 | 550 | ||
| AK225867.1 | - | 2 | 504 | |||
| AK300518.1 | - | 3 | 594 | |||
| AK301765.1 | - | 3 | 920 | |||
| AK314050.1 | - | 2 | 455 | |||
| BC035606.1 | - | 2 | 425 | 5172588 | ||

FSCN3
Genomic Map