siRNA Details
| Gene Symbol: TLX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021025.3 | NP_066305.2 | 1 | 307 | ||
| GenBank mRNA | BC017291.2 | - | 1 | 307 | 4906239 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TLX3| Gene Symbol: TLX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021025.3 | NP_066305.2 | 1 | 307 | ||
| GenBank mRNA | BC017291.2 | - | 1 | 307 | 4906239 | |