siRNA Details
| Gene Symbol: EHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014601.3 | NP_055416.2 | 2 | 625 | ||
| GenBank mRNA | AF181263.1 | - | 1 | 413 | ||
| AF454952.1 | - | 1 | 430 | |||
| AK074701.1 | - | 2 | 547 | |||
| AK296907.1 | - | 3 | 517 | |||
| AK315548.1 | - | 2 | 549 | |||
| BC014445.1 | - | 2 | 536 | 4908085 | ||

EHD2
Genomic Map