siRNA Details
| Gene Symbol: CDR2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014603.2 | NP_055418.2 | 5 | 1772 | ||
| XM_006721852.2 | XP_006721915.1 | 5 | 1730 | |||
| GenBank mRNA | AK294006.1 | - | 5 | 1772 | ||
| BC013105.2 | - | 1 | 314 | |||
| BC047534.1 | - | 5 | 1772 | 5288296 | ||
| L02867.1 | - | 5 | 1570 | |||

CDR2L
Genomic Map