siRNA Details
| Gene Symbol: CD207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015717.4 | NP_056532.4 | 3 | 453 | ||
| XM_011532874.1 | XP_011531176.1 | 4 | 641 | |||
| XM_011532875.2 | XP_011531177.1 | 5 | 658 | |||
| XM_011532876.2 | XP_011531178.1 | 5 | 657 | |||
| GenBank mRNA | AJ242859.1 | - | 3 | 453 | ||
| AK314927.1 | - | 3 | 447 | |||
| BC022278.1 | - | 3 | 446 | 4692155 | ||

CD207
Genomic Map