siRNA Details
| Gene Symbol: PODXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015720.3 | NP_056535.1 | 6, 7 | 1491 | ||
| GenBank mRNA | AF219137.1 | - | 6, 7 | 1450 | ||
| AY359096.1 | - | 6, 7 | 1454 | |||
| BC019330.1 | - | 5, 6 | 1238 | 3502164 | ||
| BC052585.1 | - | 6, 7 | 1446 | 6264322 | ||

PODXL2
Genomic Map