siRNA Details
| Gene Symbol: CHST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173982.1 | NP_001167453.1 | 3 | 999 | ||
| NM_018413.5 | NP_060883.1 | 3 | 1014 | |||
| XM_017019369.1 | XP_016874858.1 | 5 | 585 | |||
| GenBank mRNA | AB042326.1 | - | 3 | 902 | ||
| AF239820.1 | - | 3 | 665 | |||
| AJ269537.2 | - | 3 | 583 | |||
| AJ289134.1 | - | 3 | 641 | |||
| AK290923.1 | - | 3 | 955 | |||
| AK313217.1 | - | 3 | 942 | |||
| BC013315.1 | - | 4 | 811 | 4042813 | ||

CHST11
Genomic Map