siRNA Details
| Gene Symbol: DHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021044.3 | NP_066382.1 | 3 | 1456 | ||
| XM_017019380.1 | XP_016874869.1 | 3 | 1387 | |||
| XM_017019381.1 | XP_016874870.1 | 3 | 1104 | |||
| GenBank mRNA | AK313360.1 | - | 3 | 1426 | ||
| BC033507.1 | - | 3 | 1456 | 5169286 | ||
| HQ607677.1 | - | 3 | 1453 | |||
| HQ607678.1 | - | 3 | 1460 | |||

DHH
Genomic Map