siRNA Details
| Gene Symbol: TRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317747.1 | NP_001304676.1 | 5 | 647 | ||
| NM_016388.3 | NP_057472.2 | 6 | 758 | |||
| GenBank mRNA | AF161547.1 | - | 6 | 672 | ||
| AJ224878.1 | - | 6 | 657 | |||
| AJ240085.1 | - | 5 | 573 | |||
| AK291991.1 | - | 6 | 758 | |||
| BC025713.1 | - | 6 | 664 | 5227396 | ||
| BX329167.2 | - | 1 | 443 | |||
| BX415345.2 | - | 6 | 666 | |||

TRAT1
Genomic Map