siRNA Details
| Gene Symbol: VILL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015873.3 | NP_056957.3 | 16 | 2185 | ||
| XM_005265191.3 | XP_005265248.1 | 17 | 2293 | |||
| XM_005265192.4 | XP_005265249.1 | 17 | 3428 | |||
| XM_006713184.3 | XP_006713247.1 | 18 | 2293 | |||
| XM_011533770.2 | XP_011532072.1 | 17 | 2449 | |||
| GenBank mRNA | AB209574.1 | - | 2 | 3642 | ||
| BC000243.1 | - | 13 | 1721 | 3356294 | ||
| BC004300.1 | - | 17 | 2215 | 3622951 | ||
| D88154.1 | - | 12 | 1591 | |||
| KU178619.1 | - | 12 | 1589 | |||
| KU178620.1 | - | 11 | 1443 | |||

VILL
Genomic Map