siRNA Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016063.2 | NP_057147.2 | 2 | 341 | ||
| GenBank mRNA | AF151888.1 | - | 2 | 200 | ||
| AF529363.1 | - | 2 | 137 | |||
| BC001671.2 | - | 2 | 276 | |||
| BC003357.1 | - | 2 | 180 | |||
| BC066332.1 | - | 2 | 155 | 5294646 | ||
| BE743302.1 | - | 2 | 254 | |||
| DB447428.1 | - | 2 | 346 | |||

HDDC2
Genomic Map