siRNA Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016063.2 | NP_057147.2 | 4 | 567 | ||
| GenBank mRNA | AF151888.1 | - | 3 | 324 | ||
| AF529363.1 | - | 4 | 363 | |||
| BC001671.2 | - | 3 | 399 | |||
| BC003357.1 | - | 4 | 406 | |||
| BC066332.1 | - | 4 | 381 | 5294646 | ||
| BE743302.1 | - | 4 | 480 | |||

HDDC2
Genomic Map