siRNA Details
| Gene Symbol: MRPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016065.3 | NP_057149.1 | 3 | 515 | ||
| GenBank mRNA | AB049948.1 | - | 3 | 401 | ||
| AF151890.1 | - | 3 | 468 | |||
| AK025715.1 | - | 3 | 448 | |||
| AK098291.1 | - | 3 | 461 | |||
| BC021106.1 | - | 3 | 410 | 5020046 | ||

MRPS16
Genomic Map