siRNA Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297746.1 | NP_001284675.1 | 4 | 593 | ||
| NM_016076.4 | NP_057160.2 | 5 | 692 | |||
| NR_123735.1 | 5 | 687 | ||||
| NR_123736.1 | 4 | 598 | ||||
| XM_011544203.2 | XP_011542505.1 | 6 | 777 | |||
| XM_011544205.2 | XP_011542507.1 | 5 | 1343 | |||
| XM_011544206.2 | XP_011542508.1 | 5 | 416 | |||
| XM_011544207.2 | XP_011542509.1 | 4 | 1783 | |||
| GenBank mRNA | AF151904.1 | - | 5 | 484 | ||
| AF229834.1 | - | 4 | 334 | |||
| AK314664.1 | - | 5 | 662 | |||
| BC004485.2 | - | 5 | 639 | 3831754 | ||
| CB854052.1 | - | 4 | 350 | |||
| CN303012.1 | - | 5 | 598 | |||

DESI2
Genomic Map