siRNA Details
| Gene Symbol: UBXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015853.4 | NP_056937.2 | 8 | 1087 | ||
| XM_005274033.4 | XP_005274090.1 | 9 | 1000 | |||
| GenBank mRNA | BC001372.2 | - | 8 | 1025 | 3051213 | |
| BC032689.1 | - | 5 | 1807 | |||

UBXN1
Genomic Map